Learn More
Thermo Scientific™ Test Tube Racks for MaxQ™ Shaker
Mix and match these shaker test tube racks, available in a variety of sizes and arrays, to suit individual needs in the laboratory.
Brand:  Thermo Scientific™ 30185
Code : 04
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 11476358
  • £318.38 / Each
Estimated Shipment: 29-08-2024
to see stock.
Add to basket
Description
Description
Mix and match Thermo Scientific™ MaxQ™ Shaker Test Tube Racks, available in a variety of sizes and arrays, to suit individual needs in the laboratory. All racks are compatible with MaxQ shaker platforms.
Racks are available in a multitude of sizes, arrays, and colors. Compatible with MaxQ shaker platforms.
Specifications
Specifications
Test Tube Rack
4 x 5
17 to 20 mm Tubes
MaxQ™ Shaker Platform
White; 17 to 20 mm; 4 x 5 array
White
20