Learn More
Fisherbrand™ Test Tubes, Borosilicate Glass 3.3, with Beaded Rim, Medium Wall
Test tubes with round bottom are universal cylindrical laboratory glassware that are used e.g. for chemical reactions, pretreatments or for storing small amounts of liquid.
Brand:  Fisherbrand™ 90106196
 View more versions of this product
Product Code. 17901152
  • £39.45 / Pack of 100
Explore more special offers
Description
Description
  • Transferring substances and liquids into other states of aggregation and thus initiating further reactions is very important for the daily work in the laboratory. A wide variety of vessels, apparatus and equipment are used for these work. Reaction controls, test series and preliminary samples are usually carried out in test tubes due to the small amounts of substance removed.
  • Manufactured from borosilicate glass 3.3
  • Thick-walled and therefore mechanically very resistant
  • With round bottom
TRUSTED_SUSTAINABILITY
Specifications
Specifications
21 mL
Borosilicate Glass (3.3)
Non-sterile
No
Test Tube
150 mm
Cylindrical
Autoclavable
16 mm
1.2 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products