Learn More
Cole Parmer™ Testing Sieve, Brass
Serialized sieve with a test sieve certificate of compliance.
Brand: Cole Parmer™
Code : NEW
Additional Details : Weight : 0.60000kg
Description
- Brass frame, brass wire.
- Test sieve certificate.
- Equivalent to ASTM E11-09 compliance sieve.
Specifications
Brass | |
10 | |
2 mm | |
1 in. |
9 | |
8 in. | |
Brass |