Learn More
Tetra-n-butoxysilane, 97%, Thermo Scientific Chemicals
£32.16 - £95.17
Chemical Identifiers
CAS | 4766-57-8 |
---|---|
Molecular Formula | C16H36O4Si |
Molecular Weight (g/mol) | 320.545 |
MDL Number | MFCD00009432 |
InChI Key | UQMOLLPKNHFRAC-UHFFFAOYSA-N |
Synonym | tetrabutyl orthosilicate, tetrabutoxysilane, tetra-n-butoxysilane, silane, tetrabutoxy, silicon tetrabutoxide, butyl silicate buo 4si, silicic acid h4sio4 , tetrabutyl ester, butyl silicate c4h9o 4si, butyl silicate, tetrabutyl orthosilice |
PubChem CID | 78500 |
IUPAC Name | tetrabutyl silicate |
SMILES | CCCCO[Si](OCCCC)(OCCCC)OCCCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11352036
|
Thermo Scientific Alfa Aesar
L03141.14 |
25 g |
£32.16
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11362036
|
Thermo Scientific Alfa Aesar
L03141.22 |
100 g |
£95.17
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields and also in the alkylation and alkoxylation of silane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields and also in the alkylation and alkoxylation of silane.
Solubility
Not miscible or difficult to mix in water.
Notes
Moisture sensitive. Store away from oxidizing agents and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Chemical Identifiers
4766-57-8 | |
320.545 | |
UQMOLLPKNHFRAC-UHFFFAOYSA-N | |
78500 | |
CCCCO[Si](OCCCC)(OCCCC)OCCCC |
C16H36O4Si | |
MFCD00009432 | |
tetrabutyl orthosilicate, tetrabutoxysilane, tetra-n-butoxysilane, silane, tetrabutoxy, silicon tetrabutoxide, butyl silicate buo 4si, silicic acid h4sio4 , tetrabutyl ester, butyl silicate c4h9o 4si, butyl silicate, tetrabutyl orthosilice | |
tetrabutyl silicate |
Specifications
277°C | |
4766-57-8 | |
MFCD00009432 | |
135°C (275°F) | |
Not miscible or difficult to mix in water. | |
CCCCO[Si](OCCCC)(OCCCC)OCCCC | |
320.545 | |
320.55 | |
Characteristic | |
Moisture sensitive | |
Tetra-n-butoxysilane |
<-80°C | |
C16H36O4Si | |
1709274 | |
tetrabutyl orthosilicate, tetrabutoxysilane, tetra-n-butoxysilane, silane, tetrabutoxy, silicon tetrabutoxide, butyl silicate buo 4si, silicic acid h4sio4 , tetrabutyl ester, butyl silicate c4h9o 4si, butyl silicate, tetrabutyl orthosilice | |
UQMOLLPKNHFRAC-UHFFFAOYSA-N | |
tetrabutyl silicate | |
78500 | |
97% | |
1.413 | |
0.899 |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
missing translation for 'einecsNumber' : 225-305-8
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only