missing translation for 'onlineSavingsMsg'
Learn More
Tetra-n-butylammonium fluoride, 1M soln. in THF, Thermo Scientific Chemicals Product Code.: 11433254

Tetra-n-butylammonium fluoride, 1M soln. in THF, Thermo Scientific Chemicals

Product Code. 11433254
100 mL
Click to view available options
Quantity:
25 mL
100 mL
500 mL
This item is not returnable. View return policy

Product Code. 11433254

Brand: Thermo Scientific Alfa Aesar A10588.AE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetra-n-butylammonium fluoride, 1M soln. in THF is used as a reactant for the preparation of triple monoamine reuptake inhibitors as a new generation of antidepressants, oligoribonucleotides with phosphonate-modified linkages, conjugated dienoic acid esters using Suzuki coupling reactions and polyacetylenic glucosides. It plays an important role in aryl alkyl alcohols through Nozaki-Hiyama allylation catalyzed by chiral bipyridyldiol ligands and chromium trichloride. It is also used as a catalyst for the silylation reactions. In addition, it is used as a source of fluoride ion in organic solvents as well as for the deprotection of silyl ether groups.

Solubility
Miscible with terahydrofuran, acetonitrile, dimethyl sulfoxide and organic solvents.

Notes
Store in a cool place. Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents and oxygen.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 429-41-4
Molecular Formula C16H36FN
Molecular Weight (g/mol) 261.47
MDL Number MFCD00011747
InChI Key FPGGTKZVZWFYPV-UHFFFAOYSA-M
Synonym tetrabutylammonium fluoride, tbaf, tetrabutylazanium fluoride, tetrabutyl ammonium fluoride, tetra-n-butylammonium fluoride, tetrabutylamine, fluoride, n,n,n-tributylbutan-1-aminium fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride, n,n,n-tributyl-1-butanaminium fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride 1:1
PubChem CID 2724141
ChEBI CHEBI:51990
IUPAC Name tetrabutylazanium;fluoride
SMILES [F-].CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Density 0.903
Flash Point −17°C (1°F)
Linear Formula [CH3(CH2)3]4NF
Quantity 100 mL
UN Number UN2924
Beilstein 3570522
Sensitivity Hygroscopic
Solubility Information Miscible with terahydrofuran,acetonitrile,dimethyl sulfoxide and organic solvents.
Formula Weight 261.47
Concentration or Composition (by Analyte or Components) 1M soln. in THF
Chemical Name or Material Tetra-n-butylammonium fluoride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10241761 Certificate TypeCertificate of Analysis Date02/04/2024 Catalog NumberA10588.AE,A10588.AP,A10588.AC
Lot Number10241703 Certificate TypeCertificate of Analysis Date26/01/2024 Catalog NumberA10588.AE,A10588.AP,A10588.AC
Lot Number10241399 Certificate TypeCertificate of Analysis Date20/06/2022 Catalog NumberA10588.AE,A10588.AP,A10588.AC
Lot Number10233336 Certificate TypeCertificate of Analysis Date21/04/2022 Catalog NumberA10588.AE,A10588.AP,A10588.AC
Lot Number10186496 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10588.AE,A10588.AP,A10588.AC
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tetra-n-butylammonium fluoride, 1M solution in THF
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Tetrahydrofuran, Tetra-n-butylammonium fluoride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tetra-n-butylammonium fluoride, 1M soln. in THF, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.