Learn More
Tetra-n-butylammonium iodotetrachloride, 95%, Thermo Scientific Chemicals
£48.10
Chemical Identifiers
CAS | 15625-59-9 |
---|---|
Molecular Formula | C16H36Cl4IN |
Molecular Weight (g/mol) | 511.175 |
MDL Number | MFCD00051851 |
InChI Key | HBZJAVINJZUOEH-UHFFFAOYSA-N |
Synonym | tetra-n-butylammonium iodotetrachloride, 1-butanaminium, n,n,n-tributyl-, tetrachloroiodate 1-, 1-butanaminium, n,n,n-tributyl-, tetrachloroiodate 1-1:1, tetrabutylammonium ion; tetrachloro-?_-iodanuide |
PubChem CID | 73555556 |
SMILES | CCCC[N+](CCCC)(CCCC)CCCC.Cl[I-](Cl)(Cl)Cl |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
15625-59-9 | |
511.175 | |
HBZJAVINJZUOEH-UHFFFAOYSA-N | |
73555556 |
C16H36Cl4IN | |
MFCD00051851 | |
tetra-n-butylammonium iodotetrachloride, 1-butanaminium, n,n,n-tributyl-, tetrachloroiodate 1-, 1-butanaminium, n,n,n-tributyl-, tetrachloroiodate 1-1:1, tetrabutylammonium ion; tetrachloro-?_-iodanuide | |
CCCC[N+](CCCC)(CCCC)CCCC.Cl[I-](Cl)(Cl)Cl |
Specifications
15625-59-9 | |
C16H36Cl4IN | |
Light sensitive | |
HBZJAVINJZUOEH-UHFFFAOYSA-N | |
511.175 | |
511.19 | |
Tetra-n-butylammonium iodotetrachloride |
135°C to 139°C | |
MFCD00051851 | |
tetra-n-butylammonium iodotetrachloride, 1-butanaminium, n,n,n-tributyl-, tetrachloroiodate 1-, 1-butanaminium, n,n,n-tributyl-, tetrachloroiodate 1-1:1, tetrabutylammonium ion; tetrachloro-?_-iodanuide | |
CCCC[N+](CCCC)(CCCC)CCCC.Cl[I-](Cl)(Cl)Cl | |
73555556 | |
95% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.