missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylphosphonium bromide, 99%, Thermo Scientific Chemicals

Product Code. 11433940
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Tetra-n-butylphosphonium bromide is used as a phase-transfer catalyst at high temperatures, at which nitrogen analogues tend to undergo elimination e.g. as catalyst for the dealkoxycarbonylation of malonates and β-keto esters by heating in stearic acid. used as an ionic liquid solvent in the regioselective O-alkylation of C/O ambident nucleophiles. It is a synthetic reagent used in proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetra-n-butylphosphonium bromide is used as a phase-transfer catalyst at high temperatures, at which nitrogen analogues tend to undergo elimination e.g. as catalyst for the dealkoxycarbonylation of malonates and ß-keto esters by heating in stearic acid. used as an ionic liquid solvent in the regioselective O-alkylation of C/O ambident nucleophiles. It is a synthetic reagent used in proteomics research.

Solubility
Soluble in water (70 g/100 ml)

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is hygroscopic in nature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3115-68-2
Molecular Formula C16H36BrP
Molecular Weight (g/mol) 339.342
MDL Number MFCD00011853
InChI Key RKHXQBLJXBGEKF-UHFFFAOYSA-M
Synonym tetrabutylphosphonium bromide, tetra-n-butylphosphonium bromide, phosphonium, tetrabutyl-, bromide, px 4b, tetra butyl phosphonium bromide, tetrabutylphosphoniumbromide, tetrabutylphosphanium bromide, tetrabutyl phosphonium bromide, phosphonium, tetrabutyl-, bromide 1:1, acmc-1clh3
PubChem CID 76564
IUPAC Name tetrabutylphosphanium;bromide
SMILES CCCC[P+](CCCC)(CCCC)CCCC.[Br-]

Specifications

Melting Point 99°C to 103°C
Flash Point 290°C (554°F)
Linear Formula [CH3(CH2)3]4PBr
Quantity 25 g
UN Number UN3464
Beilstein 4160474
Formula Weight 339.35
Percent Purity 99%
Chemical Name or Material Tetra-n-butylphosphonium bromide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.