Learn More
Tetra-n-butyltin, 96%, Thermo Scientific Chemicals
£14.96 - £85.39
Chemical Identifiers
CAS | 1461-25-2 |
---|---|
MDL Number | MFCD00009422 |
InChI Key | AFCAKJKUYFLYFK-UHFFFAOYSA-N |
Synonym | tetrabutyltin, tetra-n-butyltin, stannane, tetrabutyl, tin, tetrabutyl, tetra-n-butylcin, tetra-n-butylcin czech, unii-qj7y5v377v, ccris 6322, tetra-normal-butyltin, wln: 4-sn-4&4&4Show More |
PubChem CID | 15098 |
IUPAC Name | tetrabutylstannane |
SMILES | CCCC[Sn](CCCC)(CCCC)CCCC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
10785801
|
Thermo Scientific Acros
137980050 |
5 g | Glass bottle | N/A | ||||||
10041630
|
Thermo Scientific Acros
137981000 |
100 g | Glass bottle | N/A | ||||||
10413415
|
Thermo Scientific Acros
137985000 |
500 g | Glass bottle | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1461-25-2 | |
AFCAKJKUYFLYFK-UHFFFAOYSA-N | |
15098 | |
CCCC[Sn](CCCC)(CCCC)CCCC |
MFCD00009422 | |
tetrabutyltin, tetra-n-butyltin, stannane, tetrabutyl, tin, tetrabutyl, tetra-n-butylcin, tetra-n-butylcin czech, unii-qj7y5v377v, ccris 6322, tetra-normal-butyltin, wln: 4-sn-4&4&4Show More | |
tetrabutylstannane |
Specifications
1461-25-2 | |
Colorless to Straw | |
145°C (10.0 mmHg) | |
Authentic | |
C16H36Sn | |
1.4725 to 1.4745 | |
1.057 | |
Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents | |
CCCC[Sn](CCCC)(CCCC)CCCC | |
347.14 | |
4 mPa.s (20°C) | |
96% | |
Tetra-n-butyltin |
-97°C | |
1.0570g/mL | |
107°C | |
95% min. (GC) | |
[CH3(CH2)3]4Sn | |
MFCD00009422 | |
tetrabutyltin, tetra-n-butyltin, stannane, tetrabutyl, tin, tetrabutyl, tetra-n-butylcin, tetra-n-butylcin czech, unii-qj7y5v377v, ccris 6322, tetra-normal-butyltin, wln: 4-sn-4&4&4 | |
AFCAKJKUYFLYFK-UHFFFAOYSA-N | |
tetrabutylstannane | |
15098 | |
347.14 | |
Liquid |
Safety and Handling
GHS H Statement:
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Causes damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 215-960-8
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.