missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-hexylammonium hydrogen sulfate, 98%, Thermo Scientific Chemicals

Product Code. 11394908
2 g
Click to view available options
Quantity:
2 g
This item is not returnable. View return policy
This item is not returnable. View return policy

 

Tetra-n-hexylammonium hydrogen sulfate is used as a lipophilic phase-transfer catalyst. It is also used in coatings, printing ink, rubber, glass, leather and cosmetics. Further, it acts as an efficient catalyst in ruthenium-catalyzed reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 32503-34-7
Melting Point 96°C to 102°C
Molecular Formula C24H52NO4S
MDL Number MFCD00037675
Quantity 2 g
Synonym tetrahexylammonium hydrogensulfate, tetrahexylammonium hydrogen sulfate, tetra-n-hexylammonium hydrogen sulfate, tetrahexylammonium hydrogensulphate, 1-hexanaminium, n,n,n-trihexyl-, sulfate 1:1, tetrahexylammonium hydrogen sulphate, tetrahexylazanium sulfate, acmc-1ailg, tetra-n-hexyl-ammonium hydrogensulphate, tetra-n-hexylammonium hydrogen sulphate
Solubility Information Slightly soluble in water.
InChI Key RULHPTADXJPDSN-UHFFFAOYSA-L
SMILES [O-]S([O-])(=O)=O.CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC
IUPAC Name hydrogen sulfate;tetrahexylazanium
Molecular Weight (g/mol) 450.74
PubChem CID 11015848
Formula Weight 451.76
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Tetra-n-hexylammonium hydrogen sulfate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tetra-n-hexylammonium hydrogen sulfate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.