Learn More
Tetra-n-propylammonium hydroxide, 40% w/w aq. soln., Thermo Scientific Chemicals
Description
Tetra-n-propylammonium hydroxide is used as an intermediate, ion exchange agent, surface-active agent and solvent. Further, it is used as an antistatic agent, detergent sanitizers, softener for textiles and paper products, emulsifying agents and pigment dispersers. It finds application as curing accelerators. In addition to this, it acts as a phase transfer catalyst in organic synthesis to enhance the reaction rate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | Colorless to Yellow |
Density | 0.99 g/mL |
Boiling Point | 102°C |
Linear Formula | (CH3CH2CH2)4NOH |
Refractive Index | 1.3716 |
Quantity | 500 g |
UN Number | UN3267 |
Beilstein | 4207983 |
Sensitivity | Air Sensitive |
Solubility Information | Miscible with strong electrolytes. |
Show More |
Safety and Handling
- Tetra-n-propylammonium hydroxide, 40% w/w aqueous solution
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : N,N,N-tripropyl-1-propanaminium hydroxide
RUO – Research Use Only