Learn More
Tetra-n-propylammonium hydroxide, 40% w/w aq. soln., Thermo Scientific Chemicals
Description
Tetra-n-propylammonium hydroxide is used as an intermediate, ion exchange agent, surface-active agent and solvent. Further, it is used as an antistatic agent, detergent sanitizers, softener for textiles and paper products, emulsifying agents and pigment dispersers. It finds application as curing accelerators. In addition to this, it acts as a phase transfer catalyst in organic synthesis to enhance the reaction rate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | Colorless to Yellow |
Density | 0.99 g/mL |
Boiling Point | 102°C |
Linear Formula | (CH3CH2CH2)4NOH |
Refractive Index | 1.3716 |
Quantity | 100 g |
UN Number | UN3267 |
Beilstein | 4207983 |
Sensitivity | Air Sensitive |
Solubility Information | Miscible with strong electrolytes. |
Show More |
RUO – Research Use Only