missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-propylammonium perruthenate(VII), 98%, Thermo Scientific Chemicals

Product Code. 11430328
Click to view available options
Quantity:
0.1 g
0.5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Tetra-n-propylammonium perruthenate a catalytic oxidant used in conjunction with N-methylmorpholine N-oxide for oxidation of primary & secondary alcohols to aldehydes and ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetra-n-propylammonium perruthenate a catalytic oxidant used in conjunction with N-methylmorpholine N-oxide for oxidation of primary & secondary alcohols to aldehydes and ketones.

Solubility
Insoluble in water.

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, reducing agents, heat, organic materials, metal powders, heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114615-82-6
Molecular Formula C12H35NO4Ru
Molecular Weight (g/mol) 358.485
MDL Number MFCD00074914
InChI Key HUCLFLGLPCVDMZ-UHFFFAOYSA-M
Synonym 1-propanaminium, n,n,n-tripropyl-, t-4-tetraoxoruthenate 1-
PubChem CID 127020979
IUPAC Name ruthenium;tetrapropylazanium;hydroxide;trihydrate
SMILES CCC[N+](CCC)(CCC)CCC.O.O.O.[OH-].[Ru]

Specifications

Melting Point 160°C (decomposition)
Quantity 0.1 g
Linear Formula (CH3(CH2)2)4NRuO4
UN Number UN1479
Formula Weight 351.43
Percent Purity 98%
Chemical Name or Material Tetra-n-propylammonium perruthenate(VII)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'