missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-propylammonium perruthenate(VII), 98%, Thermo Scientific Chemicals

Product Code. 11440328
Click to view available options
Quantity:
0.1 g
0.5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Tetra-n-propylammonium perruthenate a catalytic oxidant used in conjunction with N-methylmorpholine N-oxide for oxidation of primary & secondary alcohols to aldehydes and ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetra-n-propylammonium perruthenate a catalytic oxidant used in conjunction with N-methylmorpholine N-oxide for oxidation of primary & secondary alcohols to aldehydes and ketones.

Solubility
Insoluble in water.

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, reducing agents, heat, organic materials, metal powders, heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114615-82-6
Molecular Formula C12H35NO4Ru
Molecular Weight (g/mol) 358.485
MDL Number MFCD00074914
InChI Key HUCLFLGLPCVDMZ-UHFFFAOYSA-M
Synonym 1-propanaminium, n,n,n-tripropyl-, t-4-tetraoxoruthenate 1-
PubChem CID 127020979
IUPAC Name ruthenium;tetrapropylazanium;hydroxide;trihydrate
SMILES CCC[N+](CCC)(CCC)CCC.O.O.O.[OH-].[Ru]

Specifications

Melting Point 160°C (decomposition)
Quantity 0.5 g
Linear Formula (CH3(CH2)2)4NRuO4
UN Number UN1479
Solubility Information Insoluble in water.
Formula Weight 351.43
Percent Purity 98%
Sensitivity Hygroscopic
Chemical Name or Material Tetra-n-propylammonium perruthenate(VII)

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.