missing translation for 'onlineSavingsMsg'
Learn More
Tetraacetylethylenediamine, 90%, Thermo Scientific Chemicals Product Code.: 10454955

Tetraacetylethylenediamine, 90%, Thermo Scientific Chemicals

Product Code. 10454955
50 g, Plastic bottle
Click to view available options
Quantity:
50 g
250 g
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10454955

Brand: Thermo Scientific Acros 344590500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10543-57-4
Molecular Formula C10H16N2O4
Molecular Weight (g/mol) 228.25
MDL Number MFCD00014967
InChI Key BGRWYDHXPHLNKA-UHFFFAOYSA-N
Synonym tetraacetylethylenediamine, n,n,n',n'-tetraacetylethylenediamine, taed, tetracetylethylenediamine, acetamide, n,n'-1,2-ethanediylbis n-acetyl, n,n'-ethylenebis n-acetylacetamide, unii-p411ed0n2b, n,n'-ethane-1,2-diyl bis n-acetylacetamide, n-acetyl-n-2-diacetylamino ethyl acetamideShow MoreShow Less
PubChem CID 66347
IUPAC Name N-acetyl-N-[2-(diacetylamino)ethyl]acetamide
SMILES CC(=O)N(CCN(C(=O)C)C(=O)C)C(=O)C

Specifications

Melting Point 149°C to 154°C
Color White to Beige
Boiling Point 140°C (2.0 hPa)
Flash Point 140°C
Infrared Spectrum Authentic
Assay Percent Range 88% min
Packaging Plastic bottle
Quantity 50 g
Merck Index 15, 9158
Solubility Information Solubility in water: ca 1g/L (20°C). Other solubilities: slightly soluble in acetone,moderately soluble in chloroform,methylenechloride
Formula Weight 228.25
Percent Purity 90%
Physical Form Granular Powder
Chemical Name or Material Tetraacetylethylenediamine, 90%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465152 Certificate TypeCertificate of Analysis Date01/08/2024 Catalog Number344590500,344592500,344590000
Lot NumberA0459662 Certificate TypeCertificate of Analysis Date26/01/2024 Catalog Number344590500,344592500,344590000
Lot NumberA0456475 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog Number344590500,344592500,344590000
Lot NumberA0414459 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number344590500,344592500,344590000
Lot NumberA0403996 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number344590500,344592500,344590000
Product Identifier
  • Tetraacetylethylenediamine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tetraacetylethylenediamine, 90%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.