missing translation for 'onlineSavingsMsg'
Learn More

Tetraallyltin, 96%, Thermo Scientific Chemicals

Product Code. 11374849
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Applied as allyl transfer reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7393-43-3
Molecular Formula C12H20Sn
Molecular Weight (g/mol) 283.00
MDL Number MFCD00008637
InChI Key XJPKDRJZNZMJQM-UHFFFAOYSA-N
Synonym tetraallyltin, tetraallylstannane, tetrallylstannane, stannane, tetra-2-propenyl, stannane, tetraallyl, acmc-209osy, tetrakis prop-2-enyl stannane, stannane,tetra-2-propen-1-yl, tetrakis prop-2-en-1-yl stannane
PubChem CID 81878
IUPAC Name tetrakis(prop-2-enyl)stannane
SMILES C=CC[Sn](CC=C)(CC=C)CC=C

Specifications

Density 1.179 g/mL
Boiling Point 69°C to 70°C (1.5 mmHg)
Flash Point 75°C (167°F)
Refractive Index 1.5385
Quantity 2 g
UN Number UN2788
Sensitivity Air Sensitive
Solubility Information Not miscible in water or difficult to mix.
Formula Weight 282.98
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Tetraallyltin
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.