missing translation for 'onlineSavingsMsg'
Learn More
Tetraallyltin, 97%, Thermo Scientific™ Product Code.: 10611515

Tetraallyltin, 97%, Thermo Scientific™

Product Code. 10611515
1g; Glass bottle
Click to view available options
1g; Glass bottle
This item is not returnable. View return policy

Product Code. 10611515

Brand: Thermo Scientific Acros 382490010

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7393-43-3
Density 1.1790g/mL
Boiling Point 69.0°C to 70.0°C (1.5mmHg)
Flash Point 75°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Molecular Formula C12H20Sn
Linear Formula (CH2=CHCH2)4Sn
Refractive Index 1.5400 to 1.5420 (20°C, 589nm)
MDL Number MFCD00008637
Quantity 1g
Specific Gravity 1.179
Synonym tetraallyltin, tetraallylstannane, tetrallylstannane, stannane, tetra-2-propenyl, stannane, tetraallyl, acmc-209osy, tetrakis prop-2-enyl stannane, stannane,tetra-2-propen-1-yl, tetrakis prop-2-en-1-yl stannane
InChI Key XJPKDRJZNZMJQM-UHFFFAOYSA-N
SMILES C=CC[Sn](CC=C)(CC=C)CC=C
IUPAC Name tetrakis(prop-2-en-1-yl)stannane
Molecular Weight (g/mol) 283.00
PubChem CID 81878
Formula Weight 282.98
Percent Purity 97%
Chemical Name or Material Tetraallyltin
Show More Show Less
compliance-icons
Product Identifier
  • Tetraallyltin
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tetraallyltin, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.