missing translation for 'onlineSavingsMsg'
Learn More

Tetraamminepalladium(II) sulfate, 99.9% (metals basis), Pd 37.4% min, Thermo Scientific Chemicals

£116.58 - £2561.75

Chemical Identifiers

CAS 13601-06-4
Molecular Formula H12N4O4PdS
Molecular Weight (g/mol) 270.6
MDL Number MFCD09953444
InChI Key WSPHQZJZSJENIO-UHFFFAOYSA-L
Synonym tetraamminepalladium ii sulfate, palladium 2+ tetraamine sulfate, palladium 2+ sulfate-ammonia 1/1/4, palladium 2+ ion tetraamine sulfate
PubChem CID 18319394
IUPAC Name azane;palladium(2+);sulfate
SMILES N.N.N.N.[O-]S(=O)(=O)[O-].[Pd+2]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15426645
View Documents
Thermo Scientific Alfa Aesar
044956.03
1 g
£116.58
1g
Please to purchase this item. Need a web account? Register with us today!
15436645
View Documents
Thermo Scientific Alfa Aesar
044956.06
5 g
£589.94
5g
Please to purchase this item. Need a web account? Register with us today!
15446645
View Documents
Thermo Scientific Alfa Aesar
044956.14
25 g
£2561.75
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13601-06-4
270.6
WSPHQZJZSJENIO-UHFFFAOYSA-L
18319394
N.N.N.N.[O-]S(=O)(=O)[O-].[Pd+2]
H12N4O4PdS
MFCD09953444
tetraamminepalladium ii sulfate, palladium 2+ tetraamine sulfate, palladium 2+ sulfate-ammonia 1/1/4, palladium 2+ ion tetraamine sulfate
azane;palladium(2+);sulfate

Specifications

13601-06-4
H12N4O4PdS
tetraamminepalladium ii sulfate, palladium 2+ tetraamine sulfate, palladium 2+ sulfate-ammonia 1/1/4, palladium 2+ ion tetraamine sulfate
N.N.N.N.[O-]S(=O)(=O)[O-].[Pd+2]
270.6
270.61
Pd 37.4% min
Tetraamminepalladium (II) sulfate
Crystalline
MFCD09953444
WSPHQZJZSJENIO-UHFFFAOYSA-L
azane;palladium(2+);sulfate
18319394
99.9%
(metals basis)
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only