missing translation for 'onlineSavingsMsg'
Learn More
Tetraammineplatinum(II) nitrate, Premion™, 99.99% (metals basis), Pt 50% min, Thermo Scientific Chemicals Product Code.: 11314938

Tetraammineplatinum(II) nitrate, Premion™, 99.99% (metals basis), Pt 50% min, Thermo Scientific Chemicals

Product Code. 11314938
0.25 g
Click to view available options
Quantity:
0.25 g
1 g
5 g
This item is not returnable. View return policy

Product Code. 11314938

Brand: Thermo Scientific Alfa Aesar 043897.01

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tetraammineplatinum(II) nitrate is widely utilized as a precursor to bimetallic Pt/Zn catalysts for the selective hydrogenation of crotonaldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20634-12-2
Molecular Formula H12N6O6Pt
Molecular Weight (g/mol) 387.22
MDL Number MFCD00011624
InChI Key RBAKORNXYLGSJB-UHFFFAOYSA-N
Synonym tetraammineplatinum ii nitrate, tetraammineplatinum dinitrate, tetrammineplatinum ii nitrate, tetraammineplatinum ii nitrate 1g, tetraammineplatinum ii nitrate, premion, platinum 2+ tetraamine dinitrate, tetraammineplatinum ii nitrate solution, pt w/w cont. ptShow MoreShow Less
PubChem CID 146543
SMILES N.N.N.N.[Pt++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 262°C (decomposition)
Color White to Yellow
Physical Form Crystals
Quantity 0.25 g
Assay Percent Range 99.99% (metals basis)
Solubility Information Soluble in water.
IUPAC Name platinum(2+) tetraamine dinitrate
Formula Weight 387.21
Chemical Name or Material Tetraammineplatinum(II) nitrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY29J001 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number043897.06,043897.01,043897.03
Lot NumberY21K045 Certificate TypeCertificate of Analysis Date26/11/2024 Catalog Number043897.06,043897.01,043897.03
Lot NumberU21K026 Certificate TypeCertificate of Analysis Date06/09/2024 Catalog Number043897.06,043897.01,043897.03
Lot NumberT31K023 Certificate TypeCertificate of Analysis Date28/08/2024 Catalog Number043897.06,043897.01,043897.03
Lot NumberP22K058 Certificate TypeCertificate of Analysis Date31/03/2024 Catalog Number043897.06,043897.01,043897.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tetraammineplatinum(II) nitrate, Premion™, 99.99% (metals basis), Pt 50% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.