missing translation for 'onlineSavingsMsg'
Learn More
Tetrabenzyl pyrophosphate, 98%, Thermo Scientific Chemicals Product Code.: 11955921

Tetrabenzyl pyrophosphate, 98%, Thermo Scientific Chemicals

Product Code. 11955921
1 g
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy

Product Code. 11955921

Brand: Thermo Scientific Acros 436750010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 990-91-0
Melting Point 60.5°C to 63.0°C
Color White
Elemental Analysis 61.52 to 63.39% (C)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C28H28O7P2
Quantity 1 g
Synonym tetrabenzyl pyrophosphate, tetrabenzyl diphosphate, pyrophosphoric acid tetrabenzyl ester, tetrabenzylpyrophosphate, diphosphoric acid, tetrakis phenylmethyl ester, dibenzyl dibenzyloxyphosphoryl oxyphosphonate, benzyl pyrophosphate, acmc-209sbp, tetra benzyl pyrophosphate, tetrabenzyl diphosphate #
InChI Key NSBNXCZCLRBQTA-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)COP(=O)(OCC2=CC=CC=C2)OP(=O)(OCC3=CC=CC=C3)OCC4=CC=CC=C4
IUPAC Name dibenzyl bis(phenylmethoxy)phosphoryl phosphate
Molecular Weight (g/mol) 538.47
PubChem CID 563183
Formula Weight 538.47
Percent Purity 98%
Physical Form Crystalline Powder or Powder
Chemical Name or Material Tetrabenzyl pyrophosphate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number342425 Certificate TypeCertificate of Analysis Date01/06/2021 Catalog Number436750010
compliance-icons
Product Identifier
  • Tetrabenzyl pyrophosphate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tetrabenzyl pyrophosphate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.