missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrabromo (R) Fluorescein, Pure, Fisher Chemical™
£36.40 - £74.50
Chemical Identifiers
CAS | 17372-87-1 |
---|---|
Molecular Formula | C20H12Br4Na2O8 |
Molecular Weight (g/mol) | 745.904 |
MDL Number | 5040 |
InChI Key | MASXMTNVNASWNH-UHFFFAOYSA-L |
Synonym | eosin yellowish, eosine, bromo acid, eosine yellowish, eosine g, bromoeosine, c.i. acid red 87, sodium eosine, sodium eosinate, eosine salt free |
PubChem CID | 91886399 |
IUPAC Name | disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate |
SMILES | C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+] |
Description
Chemical Identifiers
17372-87-1 | |
745.904 | |
MASXMTNVNASWNH-UHFFFAOYSA-L | |
91886399 | |
C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+] |
C20H12Br4Na2O8 | |
5040 | |
eosin yellowish, eosine, bromo acid, eosine yellowish, eosine g, bromoeosine, c.i. acid red 87, sodium eosine, sodium eosinate, eosine salt free | |
disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate |
Specifications
17372-87-1 | |
Brown-Red | |
Glass Jar | |
5040 | |
MASXMTNVNASWNH-UHFFFAOYSA-L | |
disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate | |
91886399 | |
Powder |
300°C | |
7 to 10 | |
C20H12Br4Na2O8 | |
eosin yellowish, eosine, bromo acid, eosine yellowish, eosine g, bromoeosine, c.i. acid red 87, sodium eosine, sodium eosinate, eosine salt free | |
C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+] | |
745.904 | |
691.85g/mol |