missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium borohydride, 98%, Thermo Scientific Chemicals

Product Code. 10792251
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33725-74-5
Molecular Formula C16H40BN
Molecular Weight (g/mol) 257.31
MDL Number MFCD00012035
InChI Key GMBOFJFPOCGSOI-UHFFFAOYSA-N
Synonym tetrabutylammonium borohydride, tetra-n-butylammonium borohydride, n,n,n-tributylbutan-1-aminium tetrahydroborate, boron 1-; tetrabutylazanium, boron 1-; tetrabutylammonium
PubChem CID 9881569
IUPAC Name boron(1-);tetrabutylazanium
SMILES [BH4-].CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Melting Point 126.0°C to 131.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula [CH3(CH2)3]4NBH4
Quantity 10 g
Fieser 07,352; 10,378; 11,499
Solubility Information Solubility in water: reacts. Other solubilities: soluble in aprotic solvent
Formula Weight 257.3
Percent Purity 98%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material Tetrabutylammonium borohydride, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tetrabutylammonium borohydride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Substance which\, in contact with water\, emits flammable gases Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H261-In contact with water releases flammable gases.
Precautionary Statement
  • P223-Do not allow contact with water.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'