missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium bromide, 99+%

Product Code. 11345932 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
50 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
2.5kg
25g
500g
50g
This item is not returnable. View return policy

Product Code. 11345932

Brand: Thermo Scientific Acros 185680025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1643-19-2 | C16H36BrN | 322.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1643-19-2
Molecular Formula C16H36BrN
Molecular Weight (g/mol) 322.36
MDL Number MFCD00011633
InChI Key JRMUNVKIHCOMHV-UHFFFAOYSA-M
Synonym tetrabutylammonium bromide, tetra-n-butylammonium bromide, tbab, tetrabutylazanium bromide, tetrabutyl ammonium bromide, tetrabutyl-ammonium bromide, aliquat 100, tbabr, ipc-tba-br, unii-vjz168i98r
PubChem CID 74236
ChEBI CHEBI:51993
IUPAC Name tetrabutylazanium;bromide
SMILES CCCC[N+](CCCC)(CCCC)CCCC.[Br-]

Specifications

Melting Point 100.0°C to 104.0°C
Color Cream to White
Infrared Spectrum Authentic
Assay Percent Range 99% min. (Argentometry)
Packaging Plastic bottle
Linear Formula [CH3(CH2)3]4NBr
Quantity 2.5 kg
Fieser 04,477; 05,644; 07,353; 17,363
Solubility Information Solubility in water: 600g/L (20°C).
Formula Weight 322.36
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Tetrabutylammonium bromide, 99+%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.