missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium chloride, 95%, Tech., contains also bromide, Thermo Scientific Chemicals

Product Code. 10532391
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1112-67-0
Molecular Formula C16H36ClN
Molecular Weight (g/mol) 277.92
MDL Number MFCD00011635
InChI Key NHGXDBSUJJNIRV-UHFFFAOYSA-M
Synonym tetrabutylammonium chloride, tetrabutyl ammonium chloride, tetra-n-butylammonium chloride, tbac, tetrabutylazanium chloride, 1-butanaminium, n,n,n-tributyl-, chloride, tetrabutylammoniumchloride, 1-butanaminium, n,n,n-tributyl-, chloride 1:1, n,n,n-tributyl-1-butanaminium chloride, n,n,n-tributylbutan-1-aminium chloride
PubChem CID 70681
ChEBI CHEBI:51988
IUPAC Name tetrabutylazanium;chloride
SMILES CCCC[N+](CCCC)(CCCC)CCCC.[Cl-]

Specifications

Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (on dry substance) (Argentometry)
Packaging Glass bottle
Linear Formula [CH3(CH2)3]4NCl
Quantity 5 g
Beilstein 04, III, 292
Fieser 06,565; 07,353; 17,363
Solubility Information Solubility in water: soluble.
Formula Weight 277.92
Percent Purity 95%
Grade Technical
Physical Form Crystalline Solid
Chemical Name or Material Tetrabutylammonium chloride, Also contains bromide
Show More Show Less
compliance-icons
Product Identifier
  • Tetrabutylammonium chloride (contains also bromide)
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'