missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium fluoride, 75% solution in water, Thermo Scientific Chemicals

£29.90 - £340.00

Chemical Identifiers

CAS 429-41-4
Molecular Formula C16H36FN
Molecular Weight (g/mol) 261.47
MDL Number MFCD00011747
InChI Key FPGGTKZVZWFYPV-UHFFFAOYSA-M
Synonym tetrabutylammonium fluoride, tbaf, tetrabutylazanium fluoride, tetrabutyl ammonium fluoride, tetra-n-butylammonium fluoride, tetrabutylamine, fluoride, n,n,n-tributylbutan-1-aminium fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride, n,n,n-tributyl-1-butanaminium fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride 1:1
PubChem CID 2724141
ChEBI CHEBI:51990
SMILES [F-].CCCC[N+](CCCC)(CCCC)CCCC
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15234904
View Documents Promotion Details
Thermo Scientific Acros
451800250
25 g Glass bottle
£29.90
25g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15244904
View Documents Promotion Details
Thermo Scientific Acros
451801000
100 g Glass Bottle
£81.80
100g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15254904
View Documents Promotion Details
Thermo Scientific Acros
451805000
500 g Glass bottle
£340.00
500g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

429-41-4
261.47
FPGGTKZVZWFYPV-UHFFFAOYSA-M
2724141
[F-].CCCC[N+](CCCC)(CCCC)CCCC
C16H36FN
MFCD00011747
tetrabutylammonium fluoride, tbaf, tetrabutylazanium fluoride, tetrabutyl ammonium fluoride, tetra-n-butylammonium fluoride, tetrabutylamine, fluoride, n,n,n-tributylbutan-1-aminium fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride, n,n,n-tributyl-1-butanaminium fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride 1:1
CHEBI:51990

Specifications

429-41-4
C16H36FN
MFCD00011747
0.953
FPGGTKZVZWFYPV-UHFFFAOYSA-M
tetrabutylazanium;fluoride
2724141
261.46
Liquid
0.9530g/mL
[CH3(CH2)3]4NF
15,9332
tetrabutylammonium fluoride, tbaf, tetrabutylazanium fluoride, tetrabutyl ammonium fluoride, tetra-n-butylammonium fluoride, tetrabutylamine, fluoride, n,n,n-tributylbutan-1-aminium fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride, n,n,n-tributyl-1-butanaminium fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride 1:1
[F-].CCCC[N+](CCCC)(CCCC)CCCC
261.47
CHEBI:51990
75% in aqueous solution
Tetrabutylammonium fluoride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Causes severe skin burns and eye damage.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 207-057-2

Product Identifier
  • Tetrabutylammonium fluoride, 75% solution in water

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.

Precautionary Statement
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only