Learn More
Tetrabutylammonium hydroxide, 0.1N sol. in toluene/methanol for non-aq.titr., Thermo Scientific Chemicals
Tetrabutylammonium hydroxide, 0.1N sol. in toluene/methanol for non-aq.titr., Acros Organics, Quantity: 500mL, Packaging: Glass bottle, Colorless to Yellow, Molecular Weight: 259.47, Beilstein: 04,II,634, CAS: 2052-49-5, 108-88-3, 67-56-1, Density: 0.8500g/mL, EINECS Number: 218-147-6
Brand: Thermo Scientific Acros 123535000
78.85 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 29
Additional Details : CAS Number : 108-88-3 Weight : 0.42500kg Transport : UN number : 1993 Chem class : 3 Pack group : II
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalystSpecifications
Tetrabutylammonium hydroxide | |
Colorless to Yellow | |
4°C | |
C16H37NO | |
[CH3(CH2)3]4NOH | |
500 mL | |
05,645; 11,500; 07,332 | |
tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1 | |
VDZOOKBUILJEDG-UHFFFAOYSA-M | |
tetrabutylazanium;hydroxide | |
2723671 | |
Solution |
2052-49-5,108-88-3,67-56-1 | |
0.8500g/mL | |
Glass bottle | |
1.3775 | |
MFCD00009425 | |
04,II,634 | |
0.85 | |
Solubility in water: insoluble | |
[OH-].CCCC[N+](CCCC)(CCCC)CCCC | |
259.48 | |
259.47 |
Safety and Handling
- Tetrabutylammonium hydroxide, 0.1N sol. in toluene/methanol
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Aspiration toxicity Category 1
- Specific target organ toxicity Category 3
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after single exposure Category 1
- Serious eye damage/eye irritation Category 1
- Acute toxicity Category 4
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
- H370-Causes damage to organs.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H361d-Suspected of damaging the unborn child.
- H373-May cause damage to organs.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H225-Highly flammable liquid and vapour.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P331-Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only