missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium hydroxide 30-hydrate, 95+%, Thermo Scientific Chemicals

Catalog Number 11341513
35.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
147741-30-8
C16H97NO31
799.93
MFCD00149573
DFGIRVKFXSRUOX-UHFFFAOYSA-M
tetrabutylammonium hydroxide 30-hydrate, tetrabutylammonium ion triacontahydrate hydroxide, n,n,n-tributylbutan-1-aminium hydroxide-water 1/1/30
16218633
O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[OH-].CCCC[N+](CCCC)(CCCC)CCCC
147741-30-8
C16H97NO31
799.93
MFCD00149573
DFGIRVKFXSRUOX-UHFFFAOYSA-M
tetrabutylammonium hydroxide 30-hydrate, tetrabutylammonium ion triacontahydrate hydroxide, n,n,n-tributylbutan-1-aminium hydroxide-water 1/1/30
16218633
O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[OH-].CCCC[N+](CCCC)(CCCC)CCCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY
Melting Point 27.0°C to 30.0°C
Color White
Physical Form Crystals
Quantity 25 g
Assay Percent Range 95+%
IUPAC Name tetrabutylazanium;hydroxide;triacontahydrate
Formula Weight 799.93
Percent Purity ≥95%
Packaging Glass bottle
Chemical Name or Material Tetrabutylammonium hydroxide 30-hydrate
compliance-icons compliance-icons
Product Identifier
  • Iodine monochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H300-Fatal if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'