missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium hydroxide, 40 wt.% (1.5M) solution in water, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 176610010

Code : 29

Additional Details : CAS Number : 2052-49-5 Weight : 1.00000kg Transport : UN number : 3267 Chem class : 8 Pack group : II


 View more versions of this product

Product Code. 10782601

  • £286.00 / 1kg
In Stock
to see stock.

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Phase transfer catalyst
Specifications

Specifications

Tetrabutylammonium hydroxide
2052-49-5,7732-18-5
0.9950g/mL
Plastic bottle
[CH3(CH2)3]4NOH
1 kg
05,645; 11,500
tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1
VDZOOKBUILJEDG-UHFFFAOYSA-M
tetrabutylazanium;hydroxide
2723671
38 to 42% (Total base)
40 wt.% Solution in Water
White to Yellow
>100.0°C
C16H37NO
MFCD00009425
04, II, 634
0.995
Solubility in water: soluble.
[OH-].CCCC[N+](CCCC)(CCCC)CCCC
259.48
259.46
Crystals or Powder
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Tetrabutylammonium hydroxide, 40 wt.% solution in water

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only