missing translation for 'onlineSavingsMsg'
Learn More
Tetrabutylammonium Hydroxide, Titrant (0.4M in H2O), HPLC Grade, J.T. Baker™ Product Code.: 15568834
compliance-icons

Tetrabutylammonium Hydroxide, Titrant (0.4M in H2O), HPLC Grade, J.T. Baker™

Product Code. 15568834
500 g, HDPE Pressure Bottom Poly Bottle
Click to view available options
Quantity:
500 g
Packaging:
HDPE Pressure Bottom Poly Bottle
This item is not returnable. View return policy

Product Code. 15568834

Brand: Avantor J.T.Baker V36507

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High purity solvents for use in analytical and preparative separations

    For Use in Ion-Pair Chromatography, High Performance Liquid Chromatography

Specifications

CAS 2052-49-5
Packaging HDPE Pressure Bottom Poly Bottle
Molecular Formula C16H37NO
MDL Number MFCD00009425
Quantity 500 g
Specific Gravity 1L = 0.00kg
Synonym tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1
InChI Key VDZOOKBUILJEDG-UHFFFAOYSA-M
SMILES [OH-].CCCC[N+](CCCC)(CCCC)CCCC
IUPAC Name tetrabutylazanium;hydroxide
Molecular Weight (g/mol) 259.48
PubChem CID 2723671
Formula Weight 259.48
Grade HPLC
Chemical Name or Material Tetrabutylammonium Hydroxide, Titrant (0.4M in H2O)
Show More Show Less
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tetrabutylammonium Hydroxide, Titrant (0.4M in H2O), HPLC Grade, J.T. Baker™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.