Learn More
Tetrabutylammonium Perchlorate, Contains Max. 10% Water 98%, Thermo Scientific Chemicals
£52.46 - £530.64
Chemical Identifiers
CAS | 1923-70-2 |
---|---|
Molecular Formula | C16H36ClNO4 |
Molecular Weight (g/mol) | 341.917 |
MDL Number | MFCD00038722 |
InChI Key | KBLZDCFTQSIIOH-UHFFFAOYSA-M |
Synonym | tetrabutylammonium perchlorate, 1-butanaminium, n,n,n-tributyl-, perchlorate, tetrabutyl ammonium perchlorate, tetra-n-butylammonium perchlorate, tetrabutylammonium perchlorat, 1-butanaminium, n,n,n-tributyl-, perchlorate 1:1, acmc-1bpvk, n,n,n-tributylbutan-1-aminium perchlorate, tetrabutylammoniumperchlorate, tetrabutylazanium perchlorate |
PubChem CID | 74723 |
IUPAC Name | tetrabutylazanium;perchlorate |
SMILES | CCCC[N+](CCCC)(CCCC)CCCC.[O-]Cl(=O)(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10761062
|
Thermo Scientific Acros
420140250 |
25 g | Glass bottle |
£52.46
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10568902
|
Thermo Scientific Acros
420141000 |
100 g | Glass bottle |
£151.76
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13458558
|
Thermo Scientific Acros
420145000 |
500 g | Glass bottle |
£530.64
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent, Phase transfer catalystChemical Identifiers
1923-70-2 | |
341.917 | |
KBLZDCFTQSIIOH-UHFFFAOYSA-M | |
74723 | |
CCCC[N+](CCCC)(CCCC)CCCC.[O-]Cl(=O)(=O)=O |
C16H36ClNO4 | |
MFCD00038722 | |
tetrabutylammonium perchlorate, 1-butanaminium, n,n,n-tributyl-, perchlorate, tetrabutyl ammonium perchlorate, tetra-n-butylammonium perchlorate, tetrabutylammonium perchlorat, 1-butanaminium, n,n,n-tributyl-, perchlorate 1:1, acmc-1bpvk, n,n,n-tributylbutan-1-aminium perchlorate, tetrabutylammoniumperchlorate, tetrabutylazanium perchlorate | |
tetrabutylazanium;perchlorate |
Specifications
White | |
99% | |
(CH3(CH2)3)4NClO4 | |
tetrabutylammonium perchlorate, 1-butanaminium, n,n,n-tributyl-, perchlorate, tetrabutyl ammonium perchlorate, tetra-n-butylammonium perchlorate, tetrabutylammonium perchlorat, 1-butanaminium, n,n,n-tributyl-, perchlorate 1:1, acmc-1bpvk, n,n,n-tributylbutan-1-aminium perchlorate, tetrabutylammoniumperchlorate, tetrabutylazanium perchlorate | |
CCCC[N+](CCCC)(CCCC)CCCC.[O-]Cl(=O)(=O)=O | |
341.917 | |
341.91 | |
290°C | |
Tetrabutylammonium perchlorate, Contains Max. 10% Water |
1923-70-2 | |
C16H36ClNO4 | |
MFCD00038722 | |
KBLZDCFTQSIIOH-UHFFFAOYSA-M | |
tetrabutylazanium;perchlorate | |
74723 | |
≥97.5% | |
Crystalline Powder |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
May cause fire or explosion; strong oxidizer.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep/Store away from clothing/other combustible materials.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 217-655-5
missing translation for 'tsca' : TSCA
RUO – Research Use Only