Learn More
Tetracarbonyldi-^m-chlorodirhodium(I), Rh 50.1-52.9%, Thermo Scientific Chemicals
£151.00 - £601.00
Chemical Identifiers
CAS | 14523-22-9 |
---|---|
Molecular Formula | C4Cl2O4Rh2 |
Molecular Weight (g/mol) | 388.75 |
MDL Number | MFCD00135610 |
InChI Key | FGKDXBICTVUSPK-UHFFFAOYSA-L |
Synonym | tetracarbonyldi-mu-chlorodirhodium i, rhodium carbonyl chloride i, rhodium i carbonyl chloride, rhodium i dicarbonyl chloride dimer, di-mu-chloro-tetracarbonyldirhodium i, chlorodicarbonylrhodium i dimer, 4co.cl2rh2, bis chlorodicarbonyl rhodium, tetracarbonyldi-, i-chlorodirhodium i, tetracarbonyldi-micron-chlorodirhodium i |
IUPAC Name | bis(λ¹-rhodium(1+)) tetrakis(methanidylidyneoxidanium) dichloride |
SMILES | [Cl-].[Cl-].[Rh+].[Rh+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11366058
|
Thermo Scientific Alfa Aesar
010547.01 |
0.25 g |
£151.00
0.25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11356058
|
Thermo Scientific Alfa Aesar
010547.03 |
1 g |
£601.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used widely as a chemical for research purposes as well as it acts as a a biochemical for proteomics research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14523-22-9 | |
388.75 | |
FGKDXBICTVUSPK-UHFFFAOYSA-L | |
bis(λ¹-rhodium(1+)) tetrakis(methanidylidyneoxidanium) dichloride |
C4Cl2O4Rh2 | |
MFCD00135610 | |
tetracarbonyldi-mu-chlorodirhodium i, rhodium carbonyl chloride i, rhodium i carbonyl chloride, rhodium i dicarbonyl chloride dimer, di-mu-chloro-tetracarbonyldirhodium i, chlorodicarbonylrhodium i dimer, 4co.cl2rh2, bis chlorodicarbonyl rhodium, tetracarbonyldi-, i-chlorodirhodium i, tetracarbonyldi-micron-chlorodirhodium i | |
[Cl-].[Cl-].[Rh+].[Rh+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+] |
Specifications
14523-22-9 | |
C4Cl2O4Rh2 | |
Air and moisture sensitive | |
It is soluble in organic or non-aqueous solvent. | |
[Cl-].[Cl-].[Rh+].[Rh+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+] | |
388.75 | |
Crystalline |
121°C | |
MFCD00135610 | |
tetracarbonyldi-mu-chlorodirhodium i, rhodium carbonyl chloride i, rhodium i carbonyl chloride, rhodium i dicarbonyl chloride dimer, di-mu-chloro-tetracarbonyldirhodium i, chlorodicarbonylrhodium i dimer, 4co.cl2rh2, bis chlorodicarbonyl rhodium, tetracarbonyldi-, i-chlorodirhodium i, tetracarbonyldi-micron-chlorodirhodium i | |
FGKDXBICTVUSPK-UHFFFAOYSA-L | |
bis(λ¹-rhodium(1+)) tetrakis(methanidylidyneoxidanium) dichloride | |
388.76 | |
Tetracarbonyldi-mu-chlorodirhodium(I), Rh 50.1-52.9% |
Safety and Handling
P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
H302-H335
missing translation for 'einecsNumber' : 238-540-6
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only