Learn More
Tetrachloro-o-benzoquinone, 97%, Thermo Scientific Chemicals
£50.25 - £169.85
Chemical Identifiers
CAS | 2435-53-2 |
---|---|
Molecular Formula | C6Cl4O2 |
Molecular Weight (g/mol) | 245.86 |
MDL Number | MFCD00001646 |
InChI Key | VRGCYEIGVVTZCC-UHFFFAOYSA-N |
Synonym | o-chloranil, tetrachloro-o-benzoquinone, tetrachloro-o-quinone, 2-chloranil, tetrachloro-1,2-benzoquinone, o-chloroanil, 3,4,5,6-tetrachloro-1,2-benzoquinone, isochloranil, 3,5-cyclohexadiene-1,2-dione, 3,4,5,6-tetrachloro, ortho-chloranil |
PubChem CID | 73252 |
IUPAC Name | 3,4,5,6-tetrachlorocyclohexa-3,5-diene-1,2-dione |
SMILES | ClC1=C(Cl)C(=O)C(=O)C(Cl)=C1Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10183571
|
Thermo Scientific Acros
137990050 |
5 g | Glass bottle |
£50.25
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10051630
|
Thermo Scientific Acros
137990250 |
25 g | Glass bottle |
£169.85
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2435-53-2 | |
245.86 | |
VRGCYEIGVVTZCC-UHFFFAOYSA-N | |
73252 | |
ClC1=C(Cl)C(=O)C(=O)C(Cl)=C1Cl |
C6Cl4O2 | |
MFCD00001646 | |
o-chloranil, tetrachloro-o-benzoquinone, tetrachloro-o-quinone, 2-chloranil, tetrachloro-1,2-benzoquinone, o-chloroanil, 3,4,5,6-tetrachloro-1,2-benzoquinone, isochloranil, 3,5-cyclohexadiene-1,2-dione, 3,4,5,6-tetrachloro, ortho-chloranil | |
3,4,5,6-tetrachlorocyclohexa-3,5-diene-1,2-dione |
Specifications
2435-53-2 | |
Bordeaux | |
96% min. (GC) | |
C6Cl4(=O)2 | |
07, 602 | |
o-chloranil, tetrachloro-o-benzoquinone, tetrachloro-o-quinone, 2-chloranil, tetrachloro-1,2-benzoquinone, o-chloroanil, 3,4,5,6-tetrachloro-1,2-benzoquinone, isochloranil, 3,5-cyclohexadiene-1,2-dione, 3,4,5,6-tetrachloro, ortho-chloranil | |
VRGCYEIGVVTZCC-UHFFFAOYSA-N | |
3,4,5,6-tetrachlorocyclohexa-3,5-diene-1,2-dione | |
73252 | |
97% | |
Tetrachloro-o-benzoquinone, 97% |
125.0°C to 129.0°C | |
Authentic | |
C6Cl4O2 | |
MFCD00001646 | |
01,128; 04,76; 05,104; 07,355; 09,102; 10,85 | |
Solubility in water: insoluble. Other solubilities: 10g/L chloroform (20°C) | |
ClC1=C(Cl)C(=O)C(=O)C(Cl)=C1Cl | |
245.86 | |
245.88 | |
Fine Crystalline Powder |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 219-424-4
missing translation for 'tsca' : TSCA
- Tetrachloro-o-benzoquinone
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only