Learn More
Tetrachloro-p-benzoquinone, 99%, Thermo Scientific Chemicals
£54.70 - £185.00
Chemical Identifiers
CAS | 118-75-2 |
---|---|
Molecular Formula | C6Cl4O2 |
Molecular Weight (g/mol) | 245.86 |
MDL Number | MFCD00001594 |
InChI Key | UGNWTBMOAKPKBL-UHFFFAOYSA-N |
Synonym | chloranil, p-chloranil, tetrachloro-p-benzoquinone, tetrachloro-1,4-benzoquinone, tetrachlorobenzoquinone, spergon, coversan, reranil, vulklor, 2,3,5,6-tetrachloro-1,4-benzoquinone |
PubChem CID | 8371 |
ChEBI | CHEBI:36703 |
IUPAC Name | 2,3,5,6-tetrachlorocyclohexa-2,5-diene-1,4-dione |
SMILES | ClC1=C(Cl)C(=O)C(Cl)=C(Cl)C1=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10606642
|
Thermo Scientific Acros
213561000 |
100 g | Plastic bottle |
£54.70
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10502811
|
Thermo Scientific Acros
213562500 |
250 g | Plastic bottle |
£109.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10512811
|
Thermo Scientific Acros
213565000 |
500 g | Plastic bottle |
£185.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
118-75-2 | |
245.86 | |
UGNWTBMOAKPKBL-UHFFFAOYSA-N | |
8371 | |
2,3,5,6-tetrachlorocyclohexa-2,5-diene-1,4-dione |
C6Cl4O2 | |
MFCD00001594 | |
chloranil, p-chloranil, tetrachloro-p-benzoquinone, tetrachloro-1,4-benzoquinone, tetrachlorobenzoquinone, spergon, coversan, reranil, vulklor, 2,3,5,6-tetrachloro-1,4-benzoquinone | |
CHEBI:36703 | |
ClC1=C(Cl)C(=O)C(Cl)=C(Cl)C1=O |
Specifications
118-75-2 | |
>100°C | |
Authentic | |
C6Cl4O2 | |
MFCD00001594 | |
01,125; 02,66; 03,46; 04,75 | |
chloranil, p-chloranil, tetrachloro-p-benzoquinone, tetrachloro-1,4-benzoquinone, tetrachlorobenzoquinone, spergon, coversan, reranil, vulklor, 2,3,5,6-tetrachloro-1,4-benzoquinone | |
UGNWTBMOAKPKBL-UHFFFAOYSA-N | |
2,3,5,6-tetrachlorocyclohexa-2,5-diene-1,4-dione | |
8371 | |
245.88 | |
Liquid |
>290°C | |
0.5% max. (1 g, 105°C, 3 hrs) | |
98.5% min. (HPLC) | |
C6Cl4(=O)2 | |
07, 636 | |
15, 2078 | |
Solubility in water: insoluble. Other solubilities: soluble in ether,sparingly soluble in ccl4,cs2 and chloroform,practically insoluble in cold petroleum ether and,cold alcohol | |
ClC1=C(Cl)C(=O)C(Cl)=C(Cl)C1=O | |
245.86 | |
CHEBI:36703 | |
99% | |
Tetrachloro-p-benzoquinone |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
GHS P Statement:
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 204-274-4
missing translation for 'rtecsNumber' : DK6825000
missing translation for 'tsca' : TSCA
- Tetrachloro-p-benzoquinone
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H319-Causes serious eye irritation.
- H315-Causes skin irritation.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only