missing translation for 'onlineSavingsMsg'
Learn More

Tetrachloroethylene, ≥99.5%, Honeywell™

Product Code. 15641550
Click to view available options
Quantity:
4 x 2.5 L
6 x 1 L
Packaging:
Glass Bottle
Este artículo no se puede devolver. Vea la política de devoluciones
Este artículo no se puede devolver. Vea la política de devoluciones

≥99.5%

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 127-18-4
Molecular Formula C2Cl4
Molecular Weight (g/mol) 165.82
MDL Number MFCD00000834
InChI Key CYTYCFOTNPOANT-UHFFFAOYSA-N
Synonym tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin
PubChem CID 31373
ChEBI CHEBI:17300
IUPAC Name tetrachloroethene
SMILES ClC(Cl)=C(Cl)Cl

Especificaciones

Melting Point -22°C
Color Colorless
Density 1.623g/mL at 25°C
Boiling Point 121°C
Packaging Glass Bottle
Linear Formula CCl2=CCl2
Refractive Index n20/D 1.505(lit.)
Quantity 4 x 2.5 L
UN Number UN1897
Beilstein 1361721
Vapor Density 5.83 (vs air)
Formula Weight 165.83g/mol
Vapor Pressure 13mmHg (20°C), 19mmHg (25°C)
Percent Purity ≥99.5%
Physical Form Liquid
Chemical Name or Material Tetrachloroethylene
Mostrar más Mostrar menos
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'