missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrachloroethylene, ≥99.5%, Honeywell™
Click to view available options
Quantity:
4 x 2.5 L
6 x 1 L
Packaging:
Glass Bottle
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | -22°C |
Color | Colorless |
Density | 1.623g/mL at 25°C |
Boiling Point | 121°C |
Packaging | Glass Bottle |
Linear Formula | CCl2=CCl2 |
Refractive Index | n20/D 1.505(lit.) |
Quantity | 6 x 1 L |
UN Number | UN1897 |
Beilstein | 1361721 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'