Learn More
Tetrachloroethylene, 99%, extra pure, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Alkalinity | 0.003% max. (NaOH) |
Melting Point | -22.0°C |
Density | 1.6250g/mL |
Boiling Point | 120.0°C to 122.0°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (GC) |
Packaging | Glass Bottle |
Linear Formula | Cl2C=CCl2 |
Refractive Index | 1.505 |
Quantity | 2.5 L |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0465397 | Certificate of Analysis | 22/08/2024 | 138015000,138010010,138010025,138010100 |
A0460374 | Certificate of Analysis | 21/02/2024 | 138015000,138010010,138010025,138010100 |
A0437972 | Certificate of Analysis | 09/11/2023 | 138015000,138010010,138010025,138010100 |
A0454243 | Certificate of Analysis | 25/08/2023 | 138015000,138010010,138010025,138010100 |
A0453472 | Certificate of Analysis | 25/08/2023 | 138015000,138010010,138010025,138010100 |
Safety and Handling



- Tetrachloroethylene
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 2
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H351-Suspected of causing cancer.
- H411-Toxic to aquatic life with long lasting effects.
- H336-May cause drowsiness or dizziness.
- H315-Causes skin irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.