missing translation for 'onlineSavingsMsg'
Learn More

Tetrachloroethylene, 99%

Product Code. 11440591
Click to view available options
Quantity:
500 mL
2500 mL
Unit Size:
2500mL
500mL
This item is not returnable. View return policy

Product Code. 11440591

Brand: Thermo Scientific Alfa Aesar B20089.0F

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 127-18-4 | C2Cl4 | 165.82 g/mol

Tetrachloroethylene is used as a solvent. It is involved in dry cleaning as well as in degreasing of metal parts. It finds application as a neutrino detector. It is utilized as a paint stripper and as a spot remover in consumer products. It acts as a precursor for the manufacture of other chemicals. It plays an important role in water repellants, paint removers, printing inks, glues, sealants, polishes and lubricants. As a film-forming electrolyte additive, it is used in the manufacture of lithium ion batteries and is actively involved as an extraction solvent in the determination of oil and grease in water by FT-IR.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetrachloroethylene is used as a solvent. It is involved in dry cleaning as well as in degreasing of metal parts. It finds application as a neutrino detector. It is utilized as a paint stripper and as a spot remover in consumer products. It acts as a precursor for the manufacture of other chemicals. It plays an important role in water repellants, paint removers, printing inks, glues, sealants, polishes and lubricants. As a film-forming electrolyte additive, it is used in the manufacture of lithium ion batteries and is actively involved as an extraction solvent in the determination of oil and grease in water by FT-IR.

Solubility
Miscible with alcohol, ether, chloroform, benzene and hexane. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents, strong bases, alkali metals and aluminum.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-18-4
Molecular Formula C2Cl4
Molecular Weight (g/mol) 165.82
MDL Number MFCD00000834
InChI Key CYTYCFOTNPOANT-UHFFFAOYSA-N
Synonym tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin
PubChem CID 31373
ChEBI CHEBI:17300
SMILES ClC(Cl)=C(Cl)Cl

Specifications

Melting Point °C to 22°C
Density 1.623
Boiling Point 120°C to 121°C
Assay Percent Range 0.99
Linear Formula 2C=CCl2
Refractive Index 1.5055
Quantity 2500 mL
UN Number UN1897
Beilstein 1361721
Merck Index 14,9190
Solubility Information Miscible with alcohol,ether,chloroform,benzene and hexane. Slightly miscible with water.
IUPAC Name 1,1,2,2-tetrachloroethene
Formula Weight 165.83
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Tetrachloroethylene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tetrachloroethylene
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Tetrachloroethylene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.