missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrachloroethylene, ACS Reagent, ≥99%, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | -22°C |
Density | 1.623g/mL at 25°C |
Boiling Point | 120°C to 122°C |
Odor | Characteristic |
Packaging | Glass Bottle |
Linear Formula | CCl2=CCl2 |
Quantity | 1 L |
UN Number | UN1897 |
Beilstein | 1361721 |
Vapor Density | 5.83 (vs air) |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'