missing translation for 'onlineSavingsMsg'
Learn More

Tetrachloroethylene, Honeywell

Product Code. 15631550
Click to view available options
Quantity:
1 L
2.5 L
10 L
800 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 127-18-4
Molecular Formula C2Cl4
Molecular Weight (g/mol) 165.82
MDL Number MFCD00000834
InChI Key CYTYCFOTNPOANT-UHFFFAOYSA-N
Synonym tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin
PubChem CID 31373
ChEBI CHEBI:17300
SMILES ClC(Cl)=C(Cl)Cl

Specifications

Density 1.623g/mL at 25°C
Linear Formula CCl2=CCl2
Quantity 2.5 L
UN Number 1897
Beilstein 1361721
IUPAC Name 1,1,2,2-tetrachloroethene
Vapor Density 5.83 (vs air)
Formula Weight 165.83g/mol
Vapor Pressure 13mmHg (20°C), 19mmHg (25°C)
Percent Purity ≥99.5%
Chemical Name or Material Tetrachloroethylene
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'