missing translation for 'onlineSavingsMsg'
Learn More

Tetrachlorophthalic anhydride, 98%, Thermo Scientific Chemicals

Product Code. 10225680
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117-08-8, 118-74-1
Molecular Formula C8Cl4O3
Molecular Weight (g/mol) 285.89
MDL Number MFCD00005920
InChI Key AUHHYELHRWCWEZ-UHFFFAOYSA-N
Synonym tetrachlorophthalic anhydride, tetrathal, 4,5,6,7-tetrachloro-1,3-isobenzofurandione, niagathal, 1,3-isobenzofurandione, 4,5,6,7-tetrachloro, phthalic anhydride, tetrachloro, 4,5,6,7-tetrachloroisobenzofuran-1,3-dione, 1,3-dioxy-4,5,6,7-tetrachloroisobenzofuran, unii-76glw0lbek, ccris 6202
PubChem CID 8326
ChEBI CHEBI:59097
IUPAC Name 4,5,6,7-tetrachloro-2-benzofuran-1,3-dione
SMILES C12=C(C(=C(C(=C1Cl)Cl)Cl)Cl)C(=O)OC2=O

Specifications

Melting Point 254.0°C to 258.0°C
Color White
Boiling Point 349.0°C to 354.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Plastic bottle
Quantity 1 kg
Beilstein 17, 484
Fieser 15,300
Solubility Information Solubility in water: 0.8 mg/l (21°C). Other solubilities: hydrolysis to acid in water
Formula Weight 285.89
Percent Purity 98%
Insoluble Matter 0.5% max. (10 g in 100 ml 1 M NaOH)
Physical Form Flakes
Chemical Name or Material Tetrachlorophthalic anhydride, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'