Learn More
Thermo Scientific Chemicals Tetracycline hydrochloride, can be used as secondary standard
Tetracycline hydrochloride, CAS # 64-75-5, is a wide-spectrum antibiotic that targets both gram-negative and gram-positive bacteria.
£70.65 - £139.70
Chemical Identifiers
CAS | 64-75-5 |
---|---|
Molecular Formula | C22H25ClN2O8 |
Molecular Weight (g/mol) | 480.898 |
MDL Number | MFCD00078142 |
InChI Key | HTXDZWDXSWLLLW-FMZCEJRJSA-N |
PubChem CID | 129628373 |
IUPAC Name | (4S,4aS,5aS,6S,12aS)-4-(dimethylamino)-3,6,10,11,12a-pentahydroxy-6-methyl-1,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride |
SMILES | CC1(C2CC3C(C(=C(C(=O)C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)C(=O)N)O)N(C)C)O.Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10695272
|
Thermo Scientific Chemicals
233105000 |
500 mg | Glass Bottle |
£70.65
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10626784
|
Thermo Scientific Chemicals
233100025 |
2.5 g | Glass Bottle |
£139.70
2.50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
64-75-5 | |
480.898 | |
HTXDZWDXSWLLLW-FMZCEJRJSA-N | |
(4S,4aS,5aS,6S,12aS)-4-(dimethylamino)-3,6,10,11,12a-pentahydroxy-6-methyl-1,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride |
C22H25ClN2O8 | |
MFCD00078142 | |
129628373 | |
CC1(C2CC3C(C(=C(C(=O)C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)C(=O)N)O)N(C)C)O.Cl |
Specifications
Tetracycline Hydrochloride | |
64-75-5 | |
Authentic | |
Yellow | |
MFCD00078142 | |
15, 9341 | |
HTXDZWDXSWLLLW-FMZCEJRJSA-N | |
(4S,4aS,5aS,6S,12aS)-4-(dimethylamino)-3,6,10,11,12a-pentahydroxy-6-methyl-1,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride | |
129628373 | |
≥95 (anhydrous and solvent free), 80 wt% (as is, calculated) |
Powder | |
223.0°C | |
− 252.00 | |
C22H25ClN2O8 | |
14, IV, 2625 | |
Solubility in water: 20g/L. Other solubilities: soluble in ethanol: 5g/L, insoluble in ether, hydrocarbons and acetone | |
CC1(C2CC3C(C(=C(C(=O)C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)C(=O)N)O)N(C)C)O.Cl | |
480.898 | |
480.9 | |
Tetracycline hydrochloride |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Suspected of damaging the unborn child.
GHS P Statement:
Use personal protective equipment as required.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
LIGHT SENSITIVE
missing translation for 'einecsNumber' : 200-593-8
missing translation for 'rtecsNumber' : QI9100000
missing translation for 'tsca' : TSCA
- Tetracycline hydrochloride
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 2
Hazard Statement
- H361d-Suspected of damaging the unborn child.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.