Learn More
Tetraethoxysilane, 99+%, Thermo Scientific Chemicals
Description
As a binding agent and as a polymerization catalystTetraethoxysilane is the main precursor material for the synthesis of zeolites and silicon dioxide, which is used in semiconductor industry. It is widely used as a cross linking agent in silicon polymers and in aerogel preparations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 85°C |
Density | 0.934 g/mL |
Boiling Point | 168°C |
Flash Point | 46°C (116°F) |
Assay Percent Range | >99% |
Refractive Index | 1.382 |
Quantity | 500 g |
Beilstein | 1422225 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in ethanol and 2-propanol. |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
W21H057 | Certificate of Analysis | 12/04/2022 | 014082.18,014082.36,014082.A4,014082.57 |
D18Y003 | Certificate of Analysis | 07/12/2021 | 014082.18,014082.36,014082.A4,014082.57 |
J29Z031 | Certificate of Analysis | 07/12/2021 | 014082.18,014082.36,014082.A4,014082.57 |
R31E031 | Certificate of Analysis | 07/12/2021 | 014082.18,014082.36,014082.A4,014082.57 |
C11T002 | Certificate of Analysis | 07/12/2021 | 014082.18,014082.36,014082.A4,014082.57 |
Safety and Handling


- Tetraethoxysilane
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Specific target organ toxicity Category 3
- H226-Flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : Ethyl silicate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.