missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetraethylammonium bromide, 99+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst

Specifications
Specifications
Melting Point | 285.0°C |
Color | White to Cream |
Loss on Drying | 1.0% max. |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% min. (Argentometry) |
Packaging | Glass bottle |
Linear Formula | (C2H5)4NBr |
Quantity | 50 g |
Beilstein | 04,104 |
Fieser | 06,568; 13,46 |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0464654 | Certificate of Analysis | 15/07/2024 | 220712500,220710500,220710025,220715000 |
A0462500 | Certificate of Analysis | 01/05/2024 | 220712500,220710500,220710025,220715000 |
A0365564 | Certificate of Analysis | 13/12/2022 | 220712500,220710500,220710025,220715000 |
A0409387 | Certificate of Analysis | 13/12/2022 | |
A0430086 | Certificate of Analysis | 10/08/2022 | 220712500,220710500,220710025,220715000 |
Safety and Handling
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- H412-Harmful to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tetraethylammonium bromide, 99+%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction