missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium Hydroxide, 10% in Water, Thermo Scientific Chemicals

£19.40 - £72.40

Specifications

Chemical Name or Material Tetraethylammonium hydroxide
Color Colorless
Name Note 10% in Water
CAS 77-98-5, 7732-18-5
Boiling Point 102.0°C
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10016000
View Documents
Thermo Scientific Acros
420281000
100 g Plastic bottle
£19.40
100g
Please to purchase this item. Need a web account? Register with us today!
10026000
View Documents
Thermo Scientific Acros
420285000
500 g Plastic bottle
£72.40
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
Specifications

Specifications

Tetraethylammonium hydroxide
10% in Water
102.0°C
(C2H5)4NOH
1.0200g/mL
tetraethylammonium hydroxide, tetraethyl ammonium hydroxide, unii-ra8vu41b1f, ammonium, tetraethyl-, hydroxide, tetraethylazanium hydroxide, ethanaminium, n,n,n-triethyl-, hydroxide, ra8vu41b1f, ethanaminium, n,n,n-triethyl-, hydroxide 1:1, tetraethylammonium hydroxide solution in water, n,n,n-triethylethanaminium
LRGJRHZIDJQFCL-UHFFFAOYSA-M
tetraethylazanium;hydroxide
6509
9.5 to 10.5%
1.402
Liquid
Colorless
77-98-5, 7732-18-5
C8H21NO
MFCD00009024
15, 9346
Solubility in water: soluble.
[OH-].CC[N+](CC)(CC)CC
147.26
147.26
Plastic bottle
1.02
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Toxic to aquatic life with long lasting effects.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 201-073-3

missing translation for 'rtecsNumber' : KH3150000

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only