Learn More
Tetraethylammonium hydroxide, 25% in water, Thermo Scientific Chemicals
£12.30 - £67.64
Specifications
Chemical Name or Material | Tetraethylammonium hydroxide |
---|---|
Color | Colorless to Yellow |
Name Note | 25% in Water |
CAS | 77-98-5, 7732-18-5 |
Boiling Point | 102.0°C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10046010
|
Thermo Scientific Acros
420290250 |
25 g | Plastic bottle | N/A | |||||
10484701
|
Thermo Scientific Acros
420291000 |
100 g | Plastic bottle |
£21.61
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11965451
|
Thermo Scientific Acros
420295000 |
500 g | Plastic bottle |
£67.64
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalystSpecifications
Tetraethylammonium hydroxide | |
25% in Water | |
102.0°C | |
(C2H5)4NOH | |
1.0200g/mL | |
Solubility in water: soluble. | |
[OH-].CC[N+](CC)(CC)CC | |
147.26 | |
147.26 | |
Plastic bottle | |
1.02 |
Colorless to Yellow | |
77-98-5, 7732-18-5 | |
C8H21NO | |
MFCD00009024 | |
tetraethylammonium hydroxide, tetraethyl ammonium hydroxide, unii-ra8vu41b1f, ammonium, tetraethyl-, hydroxide, tetraethylazanium hydroxide, ethanaminium, n,n,n-triethyl-, hydroxide, ra8vu41b1f, ethanaminium, n,n,n-triethyl-, hydroxide 1:1, tetraethylammonium hydroxide solution in water, n,n,n-triethylethanaminium | |
LRGJRHZIDJQFCL-UHFFFAOYSA-M | |
tetraethylazanium;hydroxide | |
6509 | |
24 to 26% | |
1.402 | |
Solution |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Toxic to aquatic life with long lasting effects.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 201-073-3
missing translation for 'rtecsNumber' : KH3150000
missing translation for 'tsca' : TSCA
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.