missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium iodide, 98+%, Thermo Scientific Chemicals

Product Code. 11499353
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It has been used as the source of tetraethylammonium ions in pharmacological and physiological studies, but is also used in organic chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It has been used as the source of tetraethylammonium ions in pharmacological and physiological studies, but is also used in organic chemical synthesis.

Solubility
Soluble in water almost transparency.

Notes
Light Sensitive and Hygroscopic. Store in dark and away from water/moisture. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-05-3
Molecular Formula C8H20IN
Molecular Weight (g/mol) 257.16
MDL Number MFCD00011829
InChI Key UQFSVBXCNGCBBW-UHFFFAOYSA-M
Synonym tetraethylammonium iodide, tetamon iodide, tetramon j, ethanaminium, n,n,n-triethyl-, iodide, n,n,n-triethylethanaminium iodide, ammonium, tetraethyl-, iodide, teai, tetraethylammoniumiodide, tetraethylazanium iodide, tetraethyl ammonium iodide
PubChem CID 6225
IUPAC Name tetraethylazanium;iodide
SMILES [I-].CC[N+](CC)(CC)CC

Specifications

Melting Point ∼320°C (decomposition)
Quantity 250 g
Beilstein 3562649
Sensitivity Hygroscopic; Light sensitive
Solubility Information Soluble in water almost transparency.
Formula Weight 257.17
Percent Purity ≥98%
Chemical Name or Material Tetraethylammonium iodide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.