missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium perchlorate, 0.2M aqueous solution

Product Code. p-3830315 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Plastic bottle
Unit Size:
100mL
500mL
This item is not returnable. View return policy

Product Code. 10483755

Brand: Thermo Scientific Acros 379801000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2567-83-1 | C8H20ClNO4 | 229.70 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent, Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetraethylammonium perchlorate
CAS 2567-83-1, 7732-18-5
Density 1.0016g/mL
Packaging Plastic bottle
Molecular Formula C8H20ClNO4
MDL Number MFCD00041977
Quantity 100 mL
Specific Gravity 1.0016
Synonym tetraethylammonium perchlorate, ethanaminium, n,n,n-triethyl-, perchlorate, tetraethylazanium perchlorate, ethanaminium, n,n,n-triethyl-, perchlorate 1:1, ammonium, tetraethyl-, perchlorate, tetraethylammoniumperchlorate, acmc-1ccq7, ksc203k4p, tetraethyl ammonium perchlorate, tetraethylammoniumperchlorate, dry
InChI Key WGHUNMFFLAMBJD-UHFFFAOYSA-M
SMILES [O-][Cl](=O)(=O)=O.CC[N+](CC)(CC)CC
IUPAC Name tetraethylazanium;perchlorate
Molecular Weight (g/mol) 229.70
PubChem CID 2734932
Formula Weight 229.7
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.