missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetraethylene glycol dimethyl ether, 99%, Thermo Scientific Chemicals
36.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Acidity | 0.015% max. |
Melting Point | -30.0°C |
Density | 1.0090g/mL |
Boiling Point | 275.0°C to 276.0°C |
Flash Point | 141°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (GC) |
Packaging | Glass bottle |
Linear Formula | CH3(OCH2CH2)4OCH3 |
Refractive Index | 1.4310 to 1.4330 |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS08_Dangerous_Health_Hazard.png)
Product Identifier
- Tetraethylene glycol dimethyl ether
- Danger
- Reproductive toxicity Category 1B
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'