missing translation for 'onlineSavingsMsg'
Learn More
Tetraethylene glycol monooctyl ether, Thermo Scientific Chemicals Product Code.: 15454879

Tetraethylene glycol monooctyl ether, Thermo Scientific Chemicals

Product Code. 15454879
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15454879

Brand: Thermo Scientific Alfa Aesar J62496.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Reagent for solubilizing membrane proteins

Reagent for solubilizing membrane proteins

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19327-39-0
Molecular Formula C16H34O5
Molecular Weight (g/mol) 306.443
MDL Number MFCD00043033
InChI Key FEOZZFHAVXYAMB-UHFFFAOYSA-N
Synonym 3,6,9,12-tetraoxaeicosan-1-ol, 3,6,9,12-tetraoxaicosan-1-ol, tetraethylene glycol monooctyl ether, hydroxyethyloxy tri ethyloxy octane, tetraethylene glycol monoctyl ether, c8e, octyltetraglycol, octyltetraethyleneoxide, d06pvf, octyl tetraethylene glycol ether
PubChem CID 5414
ChEBI CHEBI:41323
IUPAC Name 2-[2-[2-(2-octoxyethoxy)ethoxy]ethoxy]ethanol
SMILES CCCCCCCCOCCOCCOCCOCCO

Specifications

Color Colorless
Density 0.969
Linear Formula CH3(CH2)7(OCH2CH2)4OH
Quantity 5 g
Beilstein 1781226
Formula Weight 306.44
Physical Form Liquid
Chemical Name or Material Tetraethylene glycol monooctyl ether

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberQ19K015 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberJ62496.03,J62496.14,J62496.06
Lot NumberS12J006 Certificate TypeCertificate of Analysis Date25/10/2023 Catalog NumberJ62496.03,J62496.14,J62496.06
Lot NumberT24J036 Certificate TypeCertificate of Analysis Date09/08/2023 Catalog NumberJ62496.03,J62496.14,J62496.06
Lot NumberP25I025 Certificate TypeCertificate of Analysis Date22/04/2022 Catalog NumberJ62496.03,J62496.14,J62496.06
Lot NumberU31H011 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberJ62496.03,J62496.14,J62496.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tetraethylene glycol monooctyl ether, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.