missing translation for 'onlineSavingsMsg'
Learn More

Tetraethyleneglycol monomethyl ether, 98%, Thermo Scientific Chemicals

Product Code. 10612762
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23783-42-8
Molecular Formula C9H20O5
Molecular Weight (g/mol) 208.25
MDL Number MFCD00041756
InChI Key ZNYRFEPBTVGZDN-UHFFFAOYSA-N
Synonym 2,5,8,11-tetraoxatridecan-13-ol, tetraethylene glycol monomethyl ether, tetraethyleneglycol monomethyl ether, 3,6,9,12-tetraoxatridecan-1-ol, methyl tetraglycol, tetraethyleneglycol monomethylether, mpeg4-oh, 2-2-2-2-methoxy-ethoxy-ethoxy-ethoxy-ethanol, 2-2-2-2-methoxyethoxy ethoxy ethoxy ethanol, m-peg4-alcohol
PubChem CID 90263
IUPAC Name 2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethanol
SMILES COCCOCCOCCOCCO

Specifications

Color Colorless to Yellow
Density 0.9870g/mL
Boiling Point 158°C to 160°C (5.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3O(CH2CH2O)3CH2CH2OH
Refractive Index 1.4440 to 1.446
Quantity 5 g
Specific Gravity 0.987
Formula Weight 208.25
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Tetraethyleneglycol monomethyl ether, 98%
Show More Show Less
Product Identifier
  • Tetraethyleneglycol monomethyl ether

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'