Learn More
Tetraethylenepentamine, tech., Thermo Scientific Chemicals
£23.32 - £114.57
Chemical Identifiers
CAS | 112-57-2 |
---|---|
Molecular Formula | C8H23N5 |
Molecular Weight (g/mol) | 189.31 |
MDL Number | MFCD00008168 |
InChI Key | FAGUFWYHJQFNRV-UHFFFAOYSA-N |
Synonym | tetraethylenepentamine, tetren, 1,4,7,10,13-pentaazatridecane, tetraethylene pentamine, tetraethylpentylamine, 1,11-diamino-3,6,9-triazaundecane, 3,6,9-triazaundecamethylenediamine, 3,6,9-triazaundecane-1,11-diamine, deh 26, unii-yzd1c9kq28 |
PubChem CID | 8197 |
ChEBI | CHEBI:49798 |
IUPAC Name | N'-[2-[2-(2-aminoethylamino)ethylamino]ethyl]ethane-1,2-diamine |
SMILES | NCCNCCNCCNCCN |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10205050
|
Thermo Scientific Acros
138112500 |
250 g | Glass bottle |
£23.32
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10456960
|
Thermo Scientific Acros
138115000 |
500 g | Glass bottle |
£35.58
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10005773
|
Thermo Scientific Acros
138110025 |
2.5 kg | Glass bottle |
£114.57
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
112-57-2 | |
189.31 | |
FAGUFWYHJQFNRV-UHFFFAOYSA-N | |
8197 | |
N'-[2-[2-(2-aminoethylamino)ethylamino]ethyl]ethane-1,2-diamine |
C8H23N5 | |
MFCD00008168 | |
tetraethylenepentamine, tetren, 1,4,7,10,13-pentaazatridecane, tetraethylene pentamine, tetraethylpentylamine, 1,11-diamino-3,6,9-triazaundecane, 3,6,9-triazaundecamethylenediamine, 3,6,9-triazaundecane-1,11-diamine, deh 26, unii-yzd1c9kq28 | |
CHEBI:49798 | |
NCCNCCNCCNCCN |
Specifications
112-57-2 | |
0.9900g/mL | |
139°C | |
C8H23N5 | |
1.5055 | |
0.99 | |
Solubility in water: soluble | |
NCCNCCNCCNCCN | |
189.31 | |
CHEBI:49798 | |
≥95.0% | |
Liquid |
-40°C | |
340°C | |
Authentic | |
HN(CH2CH2NHCH2CH2NH2)2 | |
MFCD00008168 | |
tetraethylenepentamine, tetren, 1,4,7,10,13-pentaazatridecane, tetraethylene pentamine, tetraethylpentylamine, 1,11-diamino-3,6,9-triazaundecane, 3,6,9-triazaundecamethylenediamine, 3,6,9-triazaundecane-1,11-diamine, deh 26, unii-yzd1c9kq28 | |
FAGUFWYHJQFNRV-UHFFFAOYSA-N | |
N'-[2-[2-(2-aminoethylamino)ethylamino]ethyl]ethane-1,2-diamine | |
8197 | |
189.3 | |
Technical | |
Tetraethylenepentamine, tech. |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Toxic in contact with skin.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously wit
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-986-2
missing translation for 'rtecsNumber' : KH8585000
missing translation for 'tsca' : TSCA
RUO – Research Use Only